siRNA Details
| Gene Symbol: Pus7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289780.1 | NP_001276709.1 | 15 | 1944 | ||
| NM_001289781.1 | NP_001276710.1 | 14 | 2277 | |||
| NM_178403.5 | NP_848490.2 | 14 | 1926 | |||
| GenBank mRNA | AK021205.1 | - | 285 | |||
| AK142271.1 | - | 14 | 1895 | |||
| AK145380.1 | - | 14 | 2277 | |||
| AK160849.1 | - | 14 | 1878 | |||
| AK173290.1 | - | 14 | 1866 | |||
| BC008544.1 | - | 14 | 1880 | 3497634 | ||
| BC010600.1 | - | 6 | 661 | |||
| BC138533.1 | - | 14 | 1768 | 8861553 | ||
| BC145316.1 | - | 15 | 1786 | 9053871 | ||

Pus7
Genomic Map