siRNA Details
| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183136.2 | NP_898959.2 | 3 | 288 | ||
| GenBank mRNA | AK021276.1 | - | 3 | 271 | ||
| AW455991.1 | - | 2 | 359 | |||
| BC048588.1 | - | 3 | 288 | 6707477 | ||
| CN843460.1 | - | 3 | 234 | |||
| DQ437329.1 | - | 3 | 270 | |||

Spink8
Genomic Map