siRNA Details
| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183136.2 | NP_898959.2 | 4, 5 | 424 | ||
| GenBank mRNA | CN843460.1 | - | 4, 5 | 370 | ||
| DQ437329.1 | - | 4, 5 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spink8| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183136.2 | NP_898959.2 | 4, 5 | 424 | ||
| GenBank mRNA | CN843460.1 | - | 4, 5 | 370 | ||
| DQ437329.1 | - | 4, 5 | 406 | |||