siRNA Details
| Gene Symbol: Troap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162506.1 | NP_001155978.1 | 8, 9 | 1013 | ||
| NM_030159.1 | NP_084435.1 | 8, 9 | 1038 | |||
| GenBank mRNA | AK050381.1 | - | 8, 9 | 1034 | ||
| AK077509.1 | - | 8, 9 | 1003 | |||
| BC080746.1 | - | 8, 9 | 998 | |||
| BC100405.1 | - | 8, 9 | 967 | |||
| BC145227.1 | - | 8, 9 | 1411 | 9053779 | ||

Troap
Genomic Map