siRNA Details
| Gene Symbol: Troap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162506.1 | NP_001155978.1 | 2 | 160 | ||
| NM_030159.1 | NP_084435.1 | 2 | 185 | |||
| GenBank mRNA | AK021408.1 | - | 2 | 172 | ||
| AK050381.1 | - | 2 | 181 | |||
| AK077509.1 | - | 2 | 150 | |||
| BC080746.1 | - | 2 | 145 | |||
| BC100405.1 | - | 2 | 114 | |||
| BC145227.1 | - | 2 | 558 | 9053779 | ||

Troap
Genomic Map