siRNA Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030166.3 | NP_084442.1 | 10 | 2222 | ||
| GenBank mRNA | AB175682.1 | - | 10 | 1820 | ||
| AK019470.1 | - | 10 | 2337 | |||
| AK030347.1 | - | 10 | 1475 | |||
| AK050075.1 | - | 10 | 1722 | |||
| AK090330.1 | - | 8 | 1545 | |||
| AK163883.1 | - | 10 | 2098 | |||
| BC133711.1 | - | 10 | 2222 | 40143107 | ||

Galnt15
Genomic Map