siRNA Details
| Gene Symbol: Rictor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030168.3 | NP_084444.3 | 18, 19 | 1688 | ||
| GenBank mRNA | AK036149.1 | - | 18, 19 | 1689 | ||
| AK082586.1 | - | 18, 19 | 1679 | |||
| AY497009.1 | - | 18, 19 | 1666 | |||
| AY540053.1 | - | 18, 19 | 1686 | |||
| BC022906.1 | - | 13, 14 | 1234 | |||
| BC120903.1 | - | 14, 15 | 1327 | 40086992 | ||
| BC120904.1 | - | 18, 19 | 1686 | 40086995 | ||

Rictor
Genomic Map