siRNA Details
| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030174.2 | NP_084450.2 | 5 | 642 | ||
| GenBank mRNA | AK047562.1 | - | 5 | 643 | ||
| AK144765.1 | - | 5 | 441 | |||
| AK154322.1 | - | 5 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mctp1| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030174.2 | NP_084450.2 | 5 | 642 | ||
| GenBank mRNA | AK047562.1 | - | 5 | 643 | ||
| AK144765.1 | - | 5 | 441 | |||
| AK154322.1 | - | 5 | 496 | |||