siRNA Details
Gene Symbol: Cfap61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175280.3 | NP_780489.2 | 2 | 182 | ||
GenBank mRNA | AK019712.1 | - | 183 | |||
AK038772.1 | - | 2 | 183 | |||
BC057361.1 | - | 2 | 150 | |||
BC058733.1 | - | 2 | 150 | |||
BC107030.1 | - | 2 | 168 | 40050496 | ||
BC107031.1 | - | 2 | 163 | 40050497 |
