siRNA Details
| Gene Symbol: Enthd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183137.2 | NP_898960.2 | 5, 6 | 441 | ||
| GenBank mRNA | AK019105.1 | - | 5, 6 | 412 | ||
| AK154660.1 | - | 5, 6 | 406 | |||
| AK181460.1 | - | 2, 3 | 199 | |||
| AK194664.1 | - | 2, 3 | 204 | |||
| AK196363.1 | - | 2, 3 | 200 | |||
| AK214680.1 | - | 2, 3 | 181 | |||
| BC094318.1 | - | 4, 5 | 930 | |||
| BC137580.1 | - | 5, 6 | 410 | 8860599 | ||
| BC144724.1 | - | 5, 6 | 410 | 9053272 | ||

Enthd2
Genomic Map