siRNA Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289613.1 | NP_001276542.1 | 10 | 1656 | ||
NM_001289616.1 | NP_001276545.1 | 11 | 1054 | |||
NM_172434.3 | NP_766022.1 | 10 | 1569 | |||
GenBank mRNA | AK019578.1 | - | 7 | 1480 | ||
AK030259.1 | - | 9 | 984 | |||
AK044506.1 | - | 11 | 1018 | |||
AY165052.1 | - | 10 | 1287 | |||
BC031740.1 | - | 7 | 817 | |||
BC057553.1 | - | 10 | 1711 | 6401356 |
