siRNA Details
| Gene Symbol: Celf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289613.1 | NP_001276542.1 | 5 | 985 | ||
| NM_001289616.1 | NP_001276545.1 | 6 | 467 | |||
| NM_001289620.1 | NP_001276549.1 | 6 | 467 | |||
| NM_172434.3 | NP_766022.1 | 5 | 982 | |||
| GenBank mRNA | AK019578.1 | - | 3 | 694 | ||
| AK030259.1 | - | 4 | 293 | |||
| AK044069.1 | - | 5 | 415 | |||
| AK044506.1 | - | 6 | 431 | |||
| AK087536.1 | - | 6 | 427 | |||
| AY165052.1 | - | 5 | 700 | |||
| BC031740.1 | - | 2 | 146 | |||
| BC057553.1 | - | 5 | 1040 | 6401356 | ||

Celf3
Genomic Map