siRNA Details
| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081253.1 | NP_001074722.1 | 2 | 1148 | ||
| GenBank mRNA | AK019714.1 | - | 2 | 1084 | ||
| AK030012.1 | - | 2 | 972 | |||
| BC098484.1 | - | 2 | 1148 | 30053208 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo43| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081253.1 | NP_001074722.1 | 2 | 1148 | ||
| GenBank mRNA | AK019714.1 | - | 2 | 1084 | ||
| AK030012.1 | - | 2 | 972 | |||
| BC098484.1 | - | 2 | 1148 | 30053208 | ||