siRNA Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030225.4 | NP_084501.1 | 5, 6 | 338 | ||
| GenBank mRNA | AK019713.2 | - | 335 | |||
| AK054053.1 | - | 5, 6 | 292 | |||
| AK149664.1 | - | 5, 6 | 314 | |||
| AK158877.1 | - | 5, 6 | 339 | |||
| AK168570.1 | - | 5, 6 | 314 | |||
| AK169943.1 | - | 5, 6 | 292 | |||
| AK202813.1 | - | 190 | ||||
| BC006702.1 | - | 5, 6 | 275 | 3583936 | ||
| CT010197.1 | - | 5, 6 | 274 | |||

Dlst
Genomic Map