siRNA Details
Gene Symbol: Agbl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048189.4 | NP_001041654.2 | 8 | 956 | ||
NM_001284190.1 | NP_001271119.1 | 8 | 956 | |||
NM_030231.3 | NP_084507.1 | 8 | 956 | |||
GenBank mRNA | AK016502.1 | - | 8 | 921 | ||
DQ867039.1 | - | 8 | 863 | |||
DQ867040.1 | - | 8 | 863 | |||
FN429928.1 | - | 8 | 782 |
