siRNA Details
| Gene Symbol: Saal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030233.1 | NP_084509.1 | 10 | 1186 | ||
| GenBank mRNA | AB489137.1 | - | 10 | 1167 | ||
| AK019878.1 | - | 10 | 1187 | |||
| AK030833.1 | - | 10 | 1192 | |||
| AK031128.1 | - | 8 | 3034 | |||
| AK033202.1 | - | 10 | 1197 | |||
| AK035874.1 | - | 9 | 3230 | |||
| AK132193.1 | - | 10 | 1187 | |||
| AK134418.1 | - | 10 | 1194 | |||
| BC019985.1 | - | 11 | 1536 | |||
| BC028473.1 | - | 5 | 600 | |||
| BC068127.1 | - | 10 | 1183 | |||
| BC087944.1 | - | 10 | 1198 | 30314036 | ||

Saal1
Genomic Map