siRNA Details
| Gene Symbol: Gm10165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512922.1 | - | 345 | |||
| CA879715.1 | - | 345 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10165| Gene Symbol: Gm10165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY512922.1 | - | 345 | |||
| CA879715.1 | - | 345 | ||||