siRNA Details
| Gene Symbol: Hdac9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271386.1 | NP_001258315.1 | 13, 14 | 1740 | ||
| NM_024124.3 | NP_077038.2 | 11, 12 | 1790 | |||
| GenBank mRNA | AF235053.1 | - | 10, 11 | 1459 | ||
| AF279371.1 | - | 9, 10 | 1327 | |||
| AF324492.1 | - | 10, 11 | 1453 | |||
| AK133334.1 | - | 13, 14 | 1740 | |||
| AK144242.1 | - | 2, 3 | 265 | |||
| BC098187.1 | - | 11, 12 | 1775 | 5363624 | ||

Hdac9
Genomic Map