siRNA Details
| Gene Symbol: Hdac9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024124.3 | NP_077038.2 | 12 | 2063 | ||
| GenBank mRNA | AF235053.1 | - | 11 | 1732 | ||
| AF279371.1 | - | 10 | 1600 | |||
| AF324492.1 | - | 11 | 1726 | |||
| AK144242.1 | - | 3 | 538 | |||
| BC098187.1 | - | 12 | 2048 | 5363624 | ||

Hdac9
Genomic Map