siRNA Details
| Gene Symbol: Lrat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023624.4 | NP_076113.1 | 1 | 621 | ||
| GenBank mRNA | AF255061.2 | - | 1 | 625 | ||
| AK004953.1 | - | 1 | 621 | |||
| BC141375.1 | - | 1 | 563 | 9055988 | ||
| BC141378.1 | - | 1 | 563 | 9055991 | ||

Lrat
Genomic Map