siRNA Details
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271768.1 | NP_001258697.1 | 4 | 413 | ||
NM_024469.2 | NP_077789.1 | 4 | 413 | |||
GenBank mRNA | AA013582.1 | - | 4 | 414 | ||
AB044090.1 | - | 4 | 333 | |||
AK137275.1 | - | 1 | 920 | |||
AK170567.1 | - | 1 | 370 | |||
BC023231.1 | - | 4 | 365 | |||
BC037742.1 | - | 4 | 373 | |||
BC051555.1 | - | 4 | 374 | |||
BE985179.1 | - | 2 | 322 |
