siRNA Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030254.3 | NP_084530.3 | 1032 | |||
| GenBank mRNA | AK028182.1 | - | 1008 | |||
| AK088616.1 | - | 1046 | ||||
| AK132766.1 | - | 1055 | ||||
| AK187688.1 | - | 19 | ||||
| BC003311.1 | - | 783 | ||||
| BC069898.1 | - | 1032 | 6489807 | |||

Tusc3
Genomic Map