siRNA Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030256.2 | NP_084532.2 | 9 | 4038 | ||
| GenBank mRNA | AB128033.1 | - | 8 | 3824 | ||
| AY296058.1 | - | 9 | 4038 | |||
| BC003321.1 | - | 1 | 258 | |||
| BC072555.1 | - | 2 | 556 | |||
| BC082304.1 | - | 8 | 4172 | 6856460 | ||

Bcl9l
Genomic Map