siRNA Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030259.1 | NP_084535.1 | 1 | 345 | ||
| GenBank mRNA | AK133760.1 | - | 1 | 477 | ||
| AK150123.1 | - | 1 | 449 | |||
| AK150252.1 | - | 1 | 448 | |||
| AK171419.1 | - | 1 | 423 | |||
| BC003324.1 | - | 1 | 345 | 3156139 | ||

Rilpl2
Genomic Map