siRNA Details
| Gene Symbol: Med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290688.1 | NP_001277617.1 | 2 | 66 | ||
| NM_020000.3 | NP_064384.2 | 2 | 66 | |||
| NM_173719.3 | NP_776067.1 | 2 | 66 | |||
| GenBank mRNA | AB041805.1 | - | 1 | 1522 | ||
| AK005422.1 | - | 2 | 49 | |||
| AK008759.1 | - | 2 | 45 | |||
| AK147032.1 | - | 2 | 52 | |||
| AK148025.1 | - | 2 | 47 | |||
| BC021870.1 | - | 2 | 37 | 4022630 | ||
| BC051683.1 | - | 2 | 44 | |||
| BC053738.1 | - | 1 | 259 | |||

Med8
Genomic Map