siRNA Details
| Gene Symbol: Pbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024954.1 | NP_001020125.1 | 3, 4 | 471 | ||
| GenBank mRNA | AJ300183.1 | - | 3, 4 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pbx4| Gene Symbol: Pbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024954.1 | NP_001020125.1 | 3, 4 | 471 | ||
| GenBank mRNA | AJ300183.1 | - | 3, 4 | 399 | ||