siRNA Details
| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 5 | 826 | ||
| GenBank mRNA | AK030596.1 | - | 3 | 396 | ||
| BC004004.1 | - | 5 | 663 | 3492768 | ||
| BC058575.1 | - | 5 | 635 | 6488611 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC004004| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 5 | 826 | ||
| GenBank mRNA | AK030596.1 | - | 3 | 396 | ||
| BC004004.1 | - | 5 | 663 | 3492768 | ||
| BC058575.1 | - | 5 | 635 | 6488611 | ||