siRNA Details
| Gene Symbol: Hcar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030701.3 | NP_109626.1 | 1 | 476 | ||
| GenBank mRNA | AJ300198.1 | - | 1 | 456 | ||
| AK150795.1 | - | 1 | 458 | |||
| AK169956.1 | - | 1 | 460 | |||
| BC120715.1 | - | 1 | 446 | 40129638 | ||
| BC125296.1 | - | 1 | 446 | 40129811 | ||

Hcar2
Genomic Map