siRNA Details
| Gene Symbol: Uck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030724.3 | NP_109649.1 | 1, 2 | 350 | ||
| GenBank mRNA | AF236636.1 | - | 1, 2 | 355 | ||
| AK051894.1 | - | 1, 2 | 120 | |||
| AK053745.1 | - | 1, 2 | 310 | |||
| AK077517.1 | - | 1, 2 | 285 | |||
| AK082837.1 | - | 1, 2 | 313 | |||
| AK155939.1 | - | 1, 2 | 314 | |||
| AK168318.1 | - | 1, 2 | 328 | |||
| BC023789.1 | - | 1, 2 | 314 | 5352192 | ||

Uck2
Genomic Map