siRNA Details
| Gene Symbol: Uck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030724.3 | NP_109649.1 | 2 | 484 | ||
| GenBank mRNA | AF236636.1 | - | 2 | 489 | ||
| AK051894.1 | - | 2 | 254 | |||
| AK053745.1 | - | 2 | 444 | |||
| AK082837.1 | - | 2 | 447 | |||
| AK155939.1 | - | 2 | 448 | |||
| AK168318.1 | - | 2 | 462 | |||
| BC023789.1 | - | 2 | 448 | 5352192 | ||

Uck2
Genomic Map