siRNA Details
| Gene Symbol: Tex13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031381.2 | NP_113558.1 | 3 | 625 | ||
| GenBank mRNA | AF285576.1 | - | 3 | 625 | ||
| AK016637.1 | - | 1 | 763 | |||
| BC116863.1 | - | 3 | 601 | 40126182 | ||
| BC119125.1 | - | 3 | 601 | 8733874 | ||

Tex13
Genomic Map