siRNA Details
| Gene Symbol: Tas1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031873.1 | NP_114079.1 | 3 | 1134 | ||
| GenBank mRNA | AF337041.1 | - | 3 | 1134 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas1r2| Gene Symbol: Tas1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031873.1 | NP_114079.1 | 3 | 1134 | ||
| GenBank mRNA | AF337041.1 | - | 3 | 1134 | ||
Set Membership: |
|