siRNA Details
| Gene Symbol: Tssk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032004.1 | NP_114393.1 | 1 | 802 | ||
| GenBank mRNA | AF329484.1 | - | 1 | 802 | ||
| AK076653.1 | - | 1 | 529 | |||
| BC049542.1 | - | 1 | 833 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tssk6| Gene Symbol: Tssk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032004.1 | NP_114393.1 | 1 | 802 | ||
| GenBank mRNA | AF329484.1 | - | 1 | 802 | ||
| AK076653.1 | - | 1 | 529 | |||
| BC049542.1 | - | 1 | 833 | |||