siRNA Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032398.2 | NP_115774.2 | 1 | 119 | ||
| GenBank mRNA | AF348403.1 | - | 1 | 90 | ||
| AF369900.1 | - | 1 | 114 | |||
| AI225679.1 | - | 1 | 119 | |||
| BC013517.1 | - | 1 | 122 | 4235957 | ||

Plvap
Genomic Map