siRNA Details
| Gene Symbol: Klhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053105.2 | NP_444335.2 | 8 | 2461 | ||
| GenBank mRNA | AF252281.1 | - | 8 | 2390 | ||
| AK031932.1 | - | 8 | 2273 | |||
| AK220397.1 | - | 8 | 1765 | |||
| BC094584.1 | - | 8 | 2461 | 30547502 | ||

Klhl1
Genomic Map