siRNA Details
| Gene Symbol: Chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053068.3 | NP_444298.1 | 1 | 130 | ||
| GenBank mRNA | AF230805.1 | - | 1 | 105 | ||
| AK010812.1 | - | 1 | 131 | |||
| AK031914.1 | - | 115 | ||||
| AK134403.1 | - | 117 | ||||
| BC016593.1 | - | 1 | 102 | |||
| BC061043.1 | - | 1 | 129 | 6773451 | ||

Chrac1
Genomic Map