siRNA Details
| Gene Symbol: Pcdhgc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033582.2 | NP_291060.1 | 1 | 1736 | ||
| GenBank mRNA | AK139349.1 | - | 1 | 474 | ||
| AK139465.1 | - | 1 | 1360 | |||
| AY013812.1 | - | 1 | 1282 | |||
| BC064808.1 | - | 1 | 1736 | 6825007 | ||

Pcdhgc4
Genomic Map