siRNA Details
| Gene Symbol: Pcdhga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033587.3 | NP_291065.3 | 1 | 2183 | ||
| GenBank mRNA | AY013798.1 | - | 1 | 2183 | ||
| BC049814.1 | - | 1 | 2309 | 6508140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga4| Gene Symbol: Pcdhga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033587.3 | NP_291065.3 | 1 | 2183 | ||
| GenBank mRNA | AY013798.1 | - | 1 | 2183 | ||
| BC049814.1 | - | 1 | 2309 | 6508140 | ||