siRNA Details
| Gene Symbol: Rnase2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053113.2 | NP_444343.2 | 2 | 386 | ||
| GenBank mRNA | BC020070.1 | - | 2 | 385 | 3982432 | |
| BC027557.1 | - | 2 | 386 | 1445853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase2a| Gene Symbol: Rnase2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053113.2 | NP_444343.2 | 2 | 386 | ||
| GenBank mRNA | BC020070.1 | - | 2 | 385 | 3982432 | |
| BC027557.1 | - | 2 | 386 | 1445853 | ||