siRNA Details
| Gene Symbol: Aff4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033565.2 | NP_291043.1 | 13, 14 | 3006 | ||
| GenBank mRNA | AF190449.1 | - | 13, 14 | 2948 | ||
| AK033163.1 | - | 3, 4 | 828 | |||
| AK053034.1 | - | 1, 2 | 36 | |||
| BC138999.1 | - | 13, 14 | 2679 | 8862021 | ||
| BC145490.1 | - | 12, 13 | 2592 | 9054053 | ||

Aff4
Genomic Map