siRNA Details
| Gene Symbol: Immp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053122.4 | NP_444352.2 | 486 | |||
| GenBank mRNA | AF359564.1 | - | 418 | |||
| AK053361.1 | - | 460 | ||||
| AK161351.1 | - | 626 | ||||
| BC061049.1 | - | 486 | ||||
| BC100557.1 | - | 438 | 5057188 | |||

Immp2l
Genomic Map