siRNA Details
Gene Symbol: Immp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053122.4 | NP_444352.2 | 564 | |||
GenBank mRNA | AF359564.1 | - | 496 | |||
AK053361.1 | - | 538 | ||||
AK161351.1 | - | 704 | ||||
BC061049.1 | - | 564 | ||||
BC100557.1 | - | 516 | 5057188 |
