siRNA Details
| Gene Symbol: Smarca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053124.2 | NP_444354.2 | 23 | 3362 | ||
| GenBank mRNA | AF325921.1 | - | 23 | 3014 | ||
| AF375046.1 | - | 23 | 3081 | |||
| AK052320.1 | - | 11 | 1315 | |||
| AK156915.1 | - | 7 | 826 | |||
| BC021922.1 | - | 11 | 1300 | |||
| BC053069.1 | - | 23 | 3362 | 6409140 | ||

Smarca5
Genomic Map