siRNA Details
Gene Symbol: Vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033509.3 | NP_277044.1 | 4 | 1018 | ||
GenBank mRNA | AF365875.1 | - | 4 | 782 | ||
AK033128.1 | - | 4 | 936 | |||
AK046036.1 | - | 4 | 1037 | |||
AK049613.1 | - | 1033 | ||||
AK129311.1 | - | 1018 | ||||
AY035370.3 | - | 4 | 1006 | |||
BC052195.1 | - | 4 | 1018 | 6509008 | ||
BC075636.1 | - | 1875 |
