siRNA Details
| Gene Symbol: Pcdhb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053147.3 | NP_444377.3 | 1 | 2482 | ||
| GenBank mRNA | AF326315.1 | - | 1 | 2354 | ||
| AK043492.1 | - | 1 | 2483 | |||
| AK143176.1 | - | 1 | 2965 | |||
| AY013784.1 | - | 1 | 2354 | |||
| BC094239.1 | - | 2 | 1955 | 30606144 | ||

Pcdhb22
Genomic Map