siRNA Details
| Gene Symbol: B3galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122993.1 | NP_001116465.1 | 4 | 721 | ||
| NM_033149.3 | NP_149161.1 | 3 | 354 | |||
| GenBank mRNA | BC047347.1 | - | 4 | 721 | ||
| BC051669.1 | - | 4 | 721 | |||
| BC057887.1 | - | 3 | 354 | 4983830 | ||

B3galt5
Genomic Map