siRNA Details
| Gene Symbol: Trim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053166.2 | NP_444396.2 | 5 | 539 | ||
| GenBank mRNA | AF396656.1 | - | 5 | 554 | ||
| AK029860.1 | - | 166 | ||||
| Gene Symbol: Gm16170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139983.1 | - | 2 | 1285 | ||

Trim7
Genomic Map