siRNA Details
| Gene Symbol: Trim34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030684.3 | NP_109609.2 | 3, 4 | 849 | ||
| GenBank mRNA | AF220139.1 | - | 3, 4 | 746 | ||
| AF220140.1 | - | 749 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim34a| Gene Symbol: Trim34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030684.3 | NP_109609.2 | 3, 4 | 849 | ||
| GenBank mRNA | AF220139.1 | - | 3, 4 | 746 | ||
| AF220140.1 | - | 749 | ||||