siRNA Details
| Gene Symbol: Mcoln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053177.1 | NP_444407.1 | 13 | 1683 | ||
| GenBank mRNA | AF302009.1 | - | 13 | 1680 | ||
| AK028385.1 | - | 13 | 1660 | |||
| AK034471.1 | - | 13 | 1694 | |||
| AK038594.1 | - | 13 | 1694 | |||
| AK049606.1 | - | 13 | 1684 | |||
| AK195153.1 | - | 2 | 52 | |||
| BC005651.1 | - | 13 | 1683 | 3257440 | ||
| BC020377.1 | - | 9 | 2125 | |||

Mcoln1
Genomic Map