siRNA Details
| Gene Symbol: Mcoln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053177.1 | NP_444407.1 | 4, 5 | 641 | ||
| GenBank mRNA | AF302009.1 | - | 4, 5 | 638 | ||
| AK028385.1 | - | 4, 5 | 618 | |||
| AK034471.1 | - | 4, 5 | 652 | |||
| AK038594.1 | - | 4, 5 | 654 | |||
| AK049606.1 | - | 4, 5 | 642 | |||
| BC005651.1 | - | 4, 5 | 641 | 3257440 | ||
| BC020377.1 | - | 3, 4 | 491 | |||

Mcoln1
Genomic Map